Silica ≥99.8%, fumed

Supplier: BeanTown Chemical

218810-100G 218810-500G
BT218810-100GEA 36.49 USD
BT218810-100G BT218810-500G
Silica ≥99.8%, fumed
Silica

CAS: 112945-52-5; MDL No: MFCD00011232; RTECS: VV7310000 Solid; Molecular Formula: SiO2; MW: 60.08 Melting Point: 1610°; Boiling Point: 2200° Density (g/mL): 2.200 Hygroscopic


  • RTECS: VV7310000
  • Hygroscopic
  • Solid

Formula: SiO₂
MW: 60.083 g/mol
Boiling Pt: 2200 °C
Melting Pt: 1610 °C
Density: 2.200 g/mL
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 112945-52-5

Order Now

Specification Test Results

Assay 99.8%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR