Germanium ≥99.9999%, pieces ≤2 cm
Supplier: BeanTown Chemical
218540-10G
218540-50G
BT218540-10GEA
189
USD
BT218540-10G
BT218540-50G
Germanium ≥99.9999%, pieces ≤2 cm
Germanium
CAS: 7440-56-4; EC No: 231-164-3; MDL No: MFCD00085310; RTECS: LY5200000
Solid, electrical resistivity: 40 ohm-cm ; Molecular Formula: Ge; MW: 72.64
Melting Point: 937°; Boiling Point: 2830°
Density (g/mL): 5.35
- RTECS: LY5200000
- Solid, electrical resistivity: 40 ohm-cm
Formula:
Ge Boiling Pt: 2830 °C Melting Pt: 937 °C Density: 5.35 g/mL |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 |
Specification Test Results
Assay | 99.9999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...