Magnesium acrylate

Supplier: BeanTown Chemical

218410-100G 218410-25G 218410-5G
BT218410-100GEA 271 USD
BT218410-100G BT218410-25G BT218410-5G
Magnesium acrylate
Magnesium acrylate

CAS: 5698-98-6; EC No: 227-177-9; MDL No: MFCD00068285 Powder; Linear Formula: (CH2=CHCOO)2Mg; Molecular Formula: C6H6MgO4 ; MW: 166.42 Melting Point: 220° (decomposes) Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: C₆H₆MgO₄
Melting Pt: >220 °C (dec.)
MDL Number: MFCD00068285
CAS Number: 5698-98-6
EINECS: 227-177-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR