Polypropylene glycol, M.W. ∼4,000
Supplier: BeanTown Chemical
Synonyms:
Poly(propylene oxide), PPG
218305-250G
218305-50G
BT218305-250GEA
89.26
USD
BT218305-250G
BT218305-50G
Polypropylene glycol, M.W. ∼4,000
Polypropylene glycol
CAS: 25322-69-4; MDL No: MFCD01779690
Liquid; Linear Formula: H[OCH(CH3)CH2]nOH; MW ≈4,000
Density (g/mL): 1.002
- Liquid
Formula:
H(OCH₂CH₂CH₂)nOH MW: 1025 Density: 1.002 g/mL |
MDL Number:
MFCD00084446 CAS Number: 25322-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...