Praseodymium ≥99% (trace metals basis excluding Ta), rods Ø 6.35 mm
Supplier: BeanTown Chemical
217640-10MM
217640-25MM
217640-50MM
BT217640-10MMEA
130
USD
BT217640-10MM
BT217640-25MM
BT217640-50MM
Praseodymium ≥99% (trace metals basis excluding Ta), rods Ø 6.35 mm
Praseodymium
CAS: 7440-10-0; EC No: 231-120-3; MDL No: MFCD00011174
Solid; Molecular Formula: Pr; MW: 140.91
Melting Point: 931°; Boiling Point: 3520°
Density (g/mL): 6.71
- Solid
Formula:
Pr MW: 140.91 Boiling Pt: 3520 °C Melting Pt: 931 °C Density: 6.71 g/mL |
MDL Number:
MFCD00011174 CAS Number: 7440-10-0 EINECS: 231-120-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...