Lansoprazole ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole, 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulfinyl)-1H-benzo[d]imidazole
216930-1G
216930-500MG
216930-5G
BT216930-1GEA
120
USD
BT216930-1G
BT216930-500MG
BT216930-5G
Lansoprazole ≥98%
Lansoprazole
CAS: 103577-45-3; MDL No: MFCD00866873; RTECS: DD9487500
Crystalline/Powder; Molecular Formula: C17H14F3N3O2; MW: 369.36
- RTECS: DD9487500
- Crystalline/Powder
Formula:
C₁₆H₁₄F₃N₃O₂S MW: 369.36 g/mol |
MDL Number:
MFCD00866873 CAS Number: 103577-45-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...