Isoniazid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide

216865-100G 216865-500G
BT216865-100GEA 36 USD
BT216865-100G BT216865-500G
Isoniazid ≥98%
Isoniazid

CAS: 54-85-3; EC No: 200-214-6; MDL No: MFCD00006426; RTECS: NS1751850 Solid; Molecular Formula: C6H7N3O2; MW: 137.14 Melting Point: 171-173° Air Sensitive


  • RTECS: NS1751850
  • Air Sensitive
  • Solid

Formula: C₆H₇N₃O
MW: 137.14 g/mol
Melting Pt: 171…173 °C
MDL Number: MFCD00006426
CAS Number: 54-85-3
EINECS: 200-214-6
Merck Index: 13,05205

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR