Magnesium ≥99.95% (trace metals basis), slug, Length 3.175 mm, Diameter 3.175 mm

Supplier: BeanTown Chemical

216490-10G 216490-50G
BT216490-10GEA 140 USD
BT216490-10G BT216490-50G
Magnesium ≥99.95% (trace metals basis), slug, Length 3.175 mm, Diameter 3.175 mm
Magnesium

CAS: 7439-95-4; EC No: 231-104-6; MDL No: MFCD00085308 Solid; Molecular Formula: Mg; MW: 24.31 Melting Point: 648°; Boiling Point: 1090° Density (g/mL): 1.74


  • Solid

Formula: Mg
Boiling Pt: 1090 °C
Melting Pt: 648 °C
Density: 1.74 g/mL
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR