1-Octanesulfonic acid sodium salt, hydrate ≥99.0% for HPLC

Supplier: BeanTown Chemical

Synonyms: Sodium 1-octansulfonate, Sodium octane-1-sulphonate

216240-100G 216240-25G 216240-5G
BT216240-100GEA 476.24 USD
BT216240-100G BT216240-25G BT216240-5G
1-Octanesulfonic acid sodium salt, hydrate ≥99.0% for HPLC
1-Octanesulphonic acid sodium salt

CAS: 5324-84-5; EC No: 226-195-4; MDL No: MFCD00007544 Crystalline; Linear Formula: CH3(CH2)7SO3Na·xH2O; Molecular Formula: C8H17NaO3S·xH2O; MW: 216.28 (anhydrous) Melting Point: 300°


  • Crystalline

Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR