Deuterium oxide ≥99.8% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: Water-D2, Heavy water

215835-250G 215835-50G
BT215835-250GEA 1030.67 USD
BT215835-250G BT215835-50G
Deuterium oxide ≥99.8% (Isotopic)
Deuterium oxide

CAS: 7789-20-0; EC No: 232-148-9; MDL No: MFCD00044636 Liquid; Molecular Formula: D2O; MW: 20.03 Melting Point: 3.8°; Boiling Point: 101.4° Density (g/mL): 1.107; Refractive Index: 1.328 Moisture Sensitive


  • Refractive index: 1.328
  • Moisture Sensitive
  • Liquid

Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.4 °C
Melting Pt: 3.8 °C
Density: 1.107 g/mL
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9

Order Now

Specification Test Results

Assay 99.8%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR