Squalene ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene

215650-100G 215650-500G
BT215650-100GEA 54.25 USD
BT215650-100G BT215650-500G
Squalene ≥98%
Squalene

CAS: 111-02-4; EC No: 203-826-1; MDL No: MFCD00008912 Liquid; Linear Formula: [(CH3)2C[=CHCH2CH2C(CH3)]2=CHCH2-]2; Molecular Formula: C30H50; MW: 410.73 Melting Point: -75°; Boiling Point: 285°/22 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.858; Refractive Index: 1.494


  • Refractive index: 1.494
  • Liquid

Formula: C₃₀H₅₀
MW: 410.73 g/mol
Boiling Pt: 285 °C/22 mmHg
Melting Pt: –75 °C
Density: 0.858 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00008912
CAS Number: 111-02-4
EINECS: 203-826-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR