Polyethylene oxide, M.W. ∼100000

Supplier: BeanTown Chemical

Synonyms: PEO

215620-100G 215620-25G 215620-500G
BT215620-100GEA 38.73 USD
BT215620-100G BT215620-25G BT215620-500G
Polyethylene oxide, M.W. ∼100000
Polyethylene oxide

CAS: 25322-68-3; EC No: 500-038-2; MDL No: MFCD00147970 Powder; Linear Formula: (-CH2CH2O-)n; MW ≈100,000 Melting Point: 65°; Boiling Point: 200° Density (g/mL): 1.13

Formula: (-CH₂CH₂O-)n
Boiling Pt: >200 °C
Melting Pt: 65 °C
Density: 1.13 g/mL
MDL Number: MFCD00147970
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR