2,6-Dichloropurine ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,6-Dichloro-9H-purine
215260-1G
215260-250MG
215260-5G
BT215260-1GEA
172
USD
BT215260-1G
BT215260-250MG
BT215260-5G
2,6-Dichloropurine ≥97%
2,6-Dichloropurine
CAS: 5451-40-1; EC No: 226-681-6; MDL No: MFCD00077725
Crystalline/Powder; Molecular Formula: C5H2Cl2N4; MW: 189.00
Melting Point: 185-195° (decomposes)
- Crystalline/Powder
Formula:
C₅H₂Cl₂N₄ MW: 189 g/mol Melting Pt: 181 (dec.) |
MDL Number:
MFCD00077725 CAS Number: 5451-40-1 EINECS: 226-681-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...