Octadecylphosphonic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: n-Octadecylphosphonic acid, ODPA

215215-1G 215215-5G
BT215215-1GEA 70 USD
BT215215-1G BT215215-5G
Octadecylphosphonic acid ≥97%
Octadecylphosphonic acid

CAS: 4724-47-4; EC No: 225-216-4; MDL No: MFCD00015838 Crystalline; Linear Formula: CH3(CH2)17P(O)(OH)2; Molecular Formula: C18H39O3P; MW: 334.48 Melting Point: 95-100°


  • Crystalline

Formula: C₁₈H₃₉O₃P
MW: 334.47 g/mol
Melting Pt: 95…100 °C
MDL Number: MFCD00015838
CAS Number: 4724-47-4
EINECS: 225-216-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR