Copper, gauze woven from wire 40 mesh Ø 0.056 mm

Supplier: BeanTown Chemical

215140-150X150 215140-300X300 215140-75X75MM
BT215140-150X1EA 145 USD
BT215140-150X1 BT215140-300X3 BT215140-75X75
Copper, gauze woven from wire 40 mesh Ø 0.056 mm
Copper

CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965 Wire Cloth; Molecular Formula: Cu; MW: 63.55 Melting Point: 1083.4°; Boiling Point: 2567° Density (g/mL): 8.94


  • Wire Cloth

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/mL
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR