3-(2-Hydroxyethyl)indole ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
215025-1G
215025-5G
BT215025-1GEA
29
USD
BT215025-1G
BT215025-5G
3-(2-Hydroxyethyl)indole ≥97%
3-(2-Hydroxyethyl)indole
CAS: 526-55-6; EC No: 208-393-2; MDL No: MFCD00005659; RTECS: KL3685000
Solid; Molecular Formula: C10H11NO ; MW: 161.20
Melting Point: 56-59°; Flash point: 255°C (491°F)
- RTECS: KL3685000
- Solid
Formula:
C₁₀H₁₁NO Melting Pt: 56…59 °C Flash Pt: 255 °C (491 °F) |
MDL Number:
MFCD00005659 CAS Number: 526-55-6 EINECS: 208-393-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...