Zinc ≥99.95% (trace metals basis), wire Ø 0.25 mm

Supplier: BeanTown Chemical

214825-25M 214825-5M
BT214825-25MEA 389 USD
BT214825-25M BT214825-5M
Zinc ≥99.95% (trace metals basis), wire Ø 0.25 mm
Zinc

CAS: 7440-66-6; EC No: 231-175-3; MDL No: MFCD00011291 Solid; Molecular Formula: Zn; MW: 65.39 Melting Point: 420°; Boiling Point: 907° Density (g/mL): 7.133


  • Solid

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.133 g/mL
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR