Octylphosphonic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: OPA, 1-Octylphosphonic acid, n-Octylphosphonic acid

214365-1G 214365-5G
BT214365-1GEA 61.78 USD
BT214365-1G BT214365-5G
Octylphosphonic acid ≥98%
Octylphosphonic acid

CAS: 4724-48-5; EC No: 225-218-5; MDL No: MFCD00015841 Crystalline; Molecular Formula: C8H19O3P; MW: 194.21 Melting Point: 93-98°


  • Crystalline

Formula: C₈H₁₉O₃P
MW: 194.21 g/mol
Melting Pt: 93…98 °C
MDL Number: MFCD00015841
CAS Number: 4724-48-5
EINECS: 225-218-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR