L(+)-Aspartic acid ≥98%
Supplier: BeanTown Chemical
213615-100G
213615-500G
BT213615-100GEA
26.88
USD
BT213615-100G
BT213615-500G
L(+)-Aspartic acid ≥98%
L(+)-Aspartic acid
CAS: 56-84-8; EC No: 200-291-6; MDL No: MFCD00002616; RTECS: CI9098500
Powder; Molecular Formula: C4H7NO4; MW: 133.10
Melting Point:
- RTECS: CI9098500
- Powder
Formula:
C₄H₇NO₄ Melting Pt: >300 °C (dec.) |
MDL Number:
MFCD00002616 CAS Number: 56-84-8 EINECS: 200-291-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...