trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl ≥97%
Supplier: BeanTown Chemical
Synonyms:
trans,trans-4-(3,4-Difluorophenyl)-4'-propylbicyclohexyl, trans,trans-4-(3,4-Difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)
212860-1G
212860-5G
BT212860-1GEA
103
USD
BT212860-1G
BT212860-5G
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl ≥97%
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl
CAS: 82832-57-3; MDL No: MFCD09838997
Powder; Molecular Formula: C21H30F2; MW: 320.47
Melting Point: 43-46°
- Powder
Formula:
C₂₁H₃₀F₂ MW: 320.468 g/mol Melting Pt: 43…46 °C |
MDL Number:
MFCD09838997 CAS Number: 82832-57-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...