Formylhydrazine ≥98%

Supplier: BeanTown Chemical

Synonyms: Formic acid hydrazide, Formohydrazide, Formic hydrazide

212190-100G 212190-25G
BT212190-100GEA 155 USD
BT212190-100G BT212190-25G
Formylhydrazine ≥98%
Formylhydrazine

CAS: 624-84-0; EC No: 210-867-9; MDL No: MFCD00007589; RTECS: LQ8615000 Flakes; Linear Formula: HCONHNH2; Molecular Formula: C4HN2O; MW: 60.06 Melting Point: 52-57°; Flash point: 112°C (234°F) Hygroscopic


  • RTECS: LQ8615000
  • Hygroscopic
  • Flakes

Formula: CH₄N₂O
MW: 60.06
Boiling Pt: 90
Melting Pt: 150
Flash Pt: 112 °C (234 °F)
MDL Number: MFCD00007589
CAS Number: 624-84-0
EINECS: 210-867-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR