1,5-Diphenylcarbazide ≥97%
Supplier: BeanTown Chemical
Synonyms:
Diphenylcarbazide, sym-Diphenylcarbazide, 1,5-Diphenylcarbohydrazide, 1,5-Diphenylcarbonohydrazide, N'',N'''-Diphenylcarbonohydrazide, sym. Diphenylcarbazide
211875-100G
211875-25G
211875-500G
BT211875-100GEA
120.04
USD
BT211875-100G
BT211875-25G
BT211875-500G
1,5-Diphenylcarbazide ≥97%
1,5-Diphenylcarbazide
CAS: 140-22-7; EC No: 205-403-7; MDL No: MFCD00003013; RTECS: FF2750000
Solid; Molecular Formula: C13H14N4O; MW: 242.28
Melting Point: 170-175°
Air Sensitive, Light Sensitive
- RTECS: FF2750000
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₁₃H₁₄N₄O MW: 242.28 g/mol Melting Pt: 170…175 °C |
MDL Number:
MFCD00003013 CAS Number: 140-22-7 EINECS: 205-403-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...