Titanium ≥99.5% (trace metals basis), wire Ø 0.81 mm. x 90cm long, annealed

Supplier: BeanTown Chemical

211625-10PC 211625-50PC
BT211625-10PCEA 92 USD
BT211625-10PC BT211625-50PC
Titanium ≥99.5% (trace metals basis), wire Ø 0.81 mm. x 90cm long, annealed
Titanium

CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264 Solid; Molecular Formula: Ti; MW: 47.87 Melting Point: 1660°; Boiling Point: 3287° Density (g/mL): 4.5


  • Solid

Formula: Ti
Boiling Pt: 3287 °C
Melting Pt: 1660 °C
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR