Sodium tetrafluoroborate ≥95%

Supplier: BeanTown Chemical

Synonyms: Sodium fluoroborate

211030-2KG 211030-500G
BT211030-2KGEA 160.79 USD
BT211030-2KG BT211030-500G
Sodium tetrafluoroborate ≥95%
Sodium tetrafluoroborate

CAS: 13755-29-8; EC No: 237-340-6; MDL No: MFCD00003515; RTECS: ED2975000 Crystalline/Granular; Linear Formula: NaBF4; MW: 109.79 Melting Point: 384° Density (g/mL): 2.47 Hygroscopic


  • RTECS: ED2975000
  • Hygroscopic
  • Granular

Formula: BF₄Na
MW: 109.79 g/mol
Melting Pt: 384 °C
Density: 2.47 g/mL
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR