Indium ≥99.995% (trace metals basis), foil, Thickness 0.25 mm
Supplier: BeanTown Chemical
144960-100X100
144960-50X50MM
BT144960-100X1EA
493
USD
BT144960-100X1
BT144960-50X50
Indium ≥99.995% (trace metals basis), foil, Thickness 0.25 mm
Indium
CAS: 7440-74-6; EC No: 231-180-0; MDL No: MFCD00134048
Solid; Molecular Formula: In; MW: 114.82
Melting Point: 156.6°; Boiling Point: 2072°
Density (g/mL): 7.3
- Solid
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.3 g/mL |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...