L(+)-Arginine ≥98%

Supplier: BeanTown Chemical

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

144805-100G 144805-25G 144805-500G
BT144805-100GEA 49.85 USD
BT144805-100G BT144805-25G BT144805-500G
L(+)-Arginine ≥98%
L(+)-Arginine

CAS: 74-79-3; EC No: 200-811-1; MDL No: MFCD00002635; RTECS: CF1934200 Crystalline/Powder; Molecular Formula: C6H14N4O2; MW: 174.20 Melting Point: 222° (decomposes) Air Sensitive


  • RTECS: CF1934200
  • Air Sensitive
  • Crystalline/Powder

Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Melting Pt: 222 °C (dec.)
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR