α-Naphtholphthalein

Supplier: BeanTown Chemical

Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one

144345-1G 144345-25G 144345-5G
BT144345-1GEA 39 USD
BT144345-1G BT144345-25G BT144345-5G
α-Naphtholphthalein
α-Naphtholphthalein

CAS: 596-01-0; EC No: 209-875-5; MDL No: MFCD00036202 Powder; Molecular Formula: C28H18O4; MW: 418.45 Melting Point: 238-240°


  • Powder

Formula: C₂₈H₁₈O₄
MW: 418.45 g/mol
Melting Pt: 238…240 °C
MDL Number: MFCD00036202
CAS Number: 596-01-0
EINECS: 209-875-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR