Zirconium ≥99% (trace metals basis excluding Hf), rods Ø 12.7 mm, annealed
Supplier: BeanTown Chemical
143905-30CM
143905-5CM
BT143905-30CMEA
317
USD
BT143905-30CM
BT143905-5CM
Zirconium ≥99% (trace metals basis excluding Hf), rods Ø 12.7 mm, annealed
Zirconium
CAS: 7440-67-7; EC No: 231-176-9; MDL No: MFCD00011303; RTECS: ZH7070000
Solid; Molecular Formula: Zr; MW: 91.22
Melting Point: 1852°; Boiling Point: 4377°
Density (g/mL): 6.5
- RTECS: ZH7070000
- Solid
Formula:
Zr Boiling Pt: 4377 °C Melting Pt: 1852 °C Density: 6.5 g/mL |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...