Methyl cellulose, 15 cP (2% H₂O)
Supplier: BeanTown Chemical
Synonyms:
Cellulose methyl ether
143895-100G
143895-2.5KG
143895-500G
BT143895-100GEA
47.23
USD
BT143895-100G
BT143895-2.5KG
BT143895-500G
Methyl cellulose, 15 cP (2% H₂O)
Methyl cellulose
CAS: 9004-67-5; MDL No: MFCD00081763
Powder
- Powder
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...