Uridine ≥99%
Supplier: BeanTown Chemical
Synonyms:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
143880-100G
143880-25G
143880-5G
BT143880-100GEA
292.35
USD
BT143880-100G
BT143880-25G
BT143880-5G
Uridine ≥99%
Uridine
CAS: 58-96-8; EC No: 200-407-5; MDL No: MFCD00006526; RTECS: YR1450000
Crystalline; Molecular Formula: C9H12N2O6; MW: 244.20
Melting Point: 163-167°
- RTECS: YR1450000
- Crystalline
Formula:
C₉H₁₂N₂O₆ MW: 244.20 g/mol Melting Pt: 163…167 °C |
MDL Number:
MFCD00006526 CAS Number: 58-96-8 EINECS: 200-407-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...