Orthoboric acid ≥99.9995% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Boron trihydroxide, Orthoboric acid, Boron hydroxide, Boric acid, Boracic acid

143385-100G 143385-25G 143385-500G
BT143385-100GEA 142 USD
BT143385-100G BT143385-25G BT143385-500G
Orthoboric acid ≥99.9995% (trace metals basis)
ortho-Boric acid

CAS: 10043-35-3; EC No: 233-139-2; MDL No: MFCD00011337; RTECS: ED4550000 Powder; Molecular Formula: H3BO3; MW: 61.83 Melting Point: 160° (decomposes)


  • RTECS: ED4550000
  • Powder

Formula: B(OH)₃
MW: 61.83 g/mol
Melting Pt: 160 °C (dec.)
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.9995%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR