5-Chloro-2-nitropyridine ≥97%
Supplier: BeanTown Chemical
143050-1G
143050-25G
143050-5G
BT143050-1GEA
57
USD
BT143050-1G
BT143050-25G
BT143050-5G
5-Chloro-2-nitropyridine ≥97%
5-Chloro-2-nitropyridine
CAS: 52092-47-4; MDL No: MFCD03092916
Solid; Molecular Formula: C5H3ClN2O2 ; MW: 158.54
Melting Point: 119-123°
- Solid
Formula:
C₅H₃ClN₂O₂ Melting Pt: 119…123 °C |
MDL Number:
MFCD03092916 CAS Number: 52092-47-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...