Tristearin (glycerol tristearate) ≥99%
Supplier: BeanTown Chemical
Synonyms:
Propane-1,2,3-triyl tristearate, Tristearin
142805-25G
142805-5G
BT142805-25GEA
687
USD
BT142805-25G
BT142805-5G
Tristearin (glycerol tristearate) ≥99%
Tristearin (Glycerol tristearate)
CAS: 555-43-1; EC No: 209-097-6; MDL No: MFCD00036230
Powder; Linear Formula: [CH3(CH2)16COOCH2]2CHOCO(CH2)16CH3; Molecular Formula: C57H110O6; MW: 891.48
Melting Point: 72°; Boiling Point: 260°
- Powder
Formula:
C₅₇H₁₁₀O₆ MW: 891.4797 g/mol Boiling Pt: 260 °C Melting Pt: 72 °C |
MDL Number:
MFCD00036230 CAS Number: 555-43-1 EINECS: 209-097-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...