Beryllium ≥99.5%, irregular lumps 3-38 mm
Supplier: BeanTown Chemical
142670-10G
142670-50G
BT142670-10GEA
86.45
USD
BT142670-10G
BT142670-50G
Beryllium ≥99.5%, irregular lumps 3-38 mm
Beryllium
CAS: 7440-41-7; EC No: 231-150-7; MDL No: MFCD00134032
Solid; Molecular Formula: Be; MW: 9.01
Melting Point: 1278°; Boiling Point: 2970°
Density (g/mL): 1.85
- Solid
Formula:
Be Boiling Pt: 2970 °C Melting Pt: 1278 °C Density: 1.85 g/mL |
MDL Number:
MFCD00134032 CAS Number: 7440-41-7 EINECS: 231-150-7 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...