Nickel ≥99% (trace metals basis), foil, annealed, Thickness 0.025 mm

Supplier: BeanTown Chemical

142280-20X30CM 142280-30X100 142280-30X500
BT142280-20X30EA 47.78 USD
BT142280-20X30 BT142280-30X10 BT142280-30X50
Nickel ≥99% (trace metals basis), foil, annealed, Thickness 0.025 mm
Nickel

CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137; RTECS: VW4725000 Solid; Molecular Formula: Ni; MW: 58.69 Melting Point: 1453°; Boiling Point: 2732° Density (g/mL): 8.9


  • RTECS: VW4725000
  • Solid

Formula: Ni
Boiling Pt: 2732 °C
Melting Pt: 1453 °C
Density: 8.9 g/mL
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR