Ferroin (1,10-phenanthrolineferrous sulfate complex) 0.025 M in water
Supplier: BeanTown Chemical
Synonyms:
Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate, Tris(1,10-phenanthroline)iron (II) sulfate, Tris(1,10-phenanthroline-N1,N10)iron sulfate
142265-250G
142265-50G
BT142265-250GEA
97
USD
BT142265-250G
BT142265-50G
Ferroin (1,10-phenanthrolineferrous sulfate complex) 0.025 M in water
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)
CAS: 14634-91-4; MDL No: MFCD00036428
Liquid; Molecular Formula: C36H24FeN6O4S ; MW: 692.52
- Liquid
Formula:
C₃₆H₂₄FeN₆SO₄ MW: 692.54 g/mol Melting Pt: 0 °C (32 °F) Density: 1 g/cm³ (8.345 lbs/gal) |
MDL Number:
MFCD00036428 CAS Number: 14634-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...