Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) ≥98%

Supplier: BeanTown Chemical

Synonyms: Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt

142235-100G 142235-2.5KG 142235-500G
BT142235-100GEA 54 USD
BT142235-100G BT142235-2.5KG BT142235-500G
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) ≥98%
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)

CAS: 140-89-6; EC No: 205-439-3; MDL No: MFCD00004931; RTECS: FG1575000 UN No: UN3342; Haz Class: 4.2; Packing Group: II Solid; Linear Formula: CH3CH2OCSSK; Molecular Formula: C3H5KOS2; MW: 160.3 Melting Point: 210° (decomposes); Flash point: 96°C (205°F) Density (g/mL): 1.558


  • RTECS: FG1575000
  • Solid

Formula: C₃H₅KOS₂
Melting Pt: 210 °C (dec.)
Density: 1.558 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00004931
CAS Number: 140-89-6
EINECS: 205-439-3
ADR: 4.2,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR