Zirconium ≥99.5% (trace metals basis excluding Hf), slug, Length 6.35 mm, Diameter 6.35 mm
Supplier: BeanTown Chemical
141395-100G
141395-25G
BT141395-100GEA
142.91
USD
BT141395-100G
BT141395-25G
Zirconium ≥99.5% (trace metals basis excluding Hf), slug, Length 6.35 mm, Diameter 6.35 mm
Zirconium
CAS: 7440-67-7; EC No: 231-176-9; MDL No: MFCD00011303
Solid; Molecular Formula: Zr; MW: 91.22
Melting Point: 1852°; Boiling Point: 4377°
Density (g/mL): 6.5
- Solid
Formula:
Zr Boiling Pt: 4377 °C Melting Pt: 1852 °C Density: 6.5 g/mL |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...