1-Aminocyclopropanecarboxylic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-Amino-1-cyclopropanecarboxylic acid
141320-1G
141320-250MG
BT141320-1GEA
145.05
USD
BT141320-1G
BT141320-250MG
1-Aminocyclopropanecarboxylic acid ≥98%
1-Aminocyclopropanecarboxylic acid
CAS: 22059-21-8; EC No: 217-162-5; MDL No: MFCD00009944; RTECS: GZ1110000
Crystalline/Powder; Molecular Formula: C4H7NO2 ; MW: 101.10
Melting Point: 229-231°
- RTECS: GZ1110000
- Crystalline/Powder
Formula:
C₄H₇NO₂ MW: 101.11 g/mol Melting Pt: 229…231 °C |
MDL Number:
MFCD00009944 CAS Number: 22059-21-8 EINECS: 217-162-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...