Isosorbide dimethyl ether ≥98%
Supplier: BeanTown Chemical
Synonyms:
(3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
140735-100G
140735-25G
BT140735-100GEA
180
USD
BT140735-100G
BT140735-25G
Isosorbide dimethyl ether ≥98%
Isosorbide dimethyl ether
CAS: 5306-85-4; EC No: 226-159-8; MDL No: MFCD00064828
Liquid; Molecular Formula: C8H14O4; MW: 174.20
Boiling Point: 93-95°/0.1 mmHg; Flash point: 108°C (226°F)
Density (g/mL): 1.15; Refractive Index: 1.462
- Refractive index: 1.462
- Liquid
Formula:
C₈H₁₄O₄ Boiling Pt: 93…95 °C/0.1 mmHg Density: 1.15 g/mL Flash Pt: 108 °C (226 °F) |
MDL Number:
MFCD00064828 CAS Number: 5306-85-4 EINECS: 226-159-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...