Platinum : rhodium (87:13) (w/w), wire 0.5 mm dia. for ISA type R standard grade thermocouple
Supplier: BeanTown Chemical
140695-100CM
140695-25CM
BT140695-100CMEA
891
USD
BT140695-100CM
BT140695-25CM
Platinum : rhodium (87:13) (w/w), wire 0.5 mm dia. for ISA type R standard grade thermocouple
Platinum : rhodium
MDL No: MFCD00198186
Solid; Formula: Pt:Rh; 87:13 wt%
- Solid