Copper ≥99.9999% (trace metals basis), slug, Length 3.175 mm, Diameter 3.175 mm
Supplier: BeanTown Chemical
140560-10G
140560-50G
BT140560-10GEA
57
USD
BT140560-10G
BT140560-50G
Copper ≥99.9999% (trace metals basis), slug, Length 3.175 mm, Diameter 3.175 mm
Copper
CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965
Solid; Molecular Formula: Cu; MW: 63.55
Melting Point: 1083.4°; Boiling Point: 2567°
Density (g/mL): 8.94
- Solid
Formula:
Cu Boiling Pt: 2567 °C Melting Pt: 1083.4 °C Density: 8.94 g/mL |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 |
Specification Test Results
Assay | 99.9999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...