Aluminium oxide, regular desiccant

Supplier: BeanTown Chemical

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

140385-100G 140385-2KG 140385-500G
BT140385-100GEA 27.98 USD
BT140385-100G BT140385-2KG BT140385-500G
Aluminium oxide, regular desiccant
Aluminium oxide

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 3.2mm Spheres; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • 3.2mm Spheres

Formula: Al₂O₃
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR