Arbutin ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Hydroxyphenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol, Hydroquinone-beta-D-glucoside
140205-1G
140205-5G
BT140205-1GEA
26
USD
BT140205-1G
BT140205-5G
Arbutin ≥98%
Arbutin
CAS: 497-76-7; EC No: 207-850-3; MDL No: MFCD00016915; RTECS: CE8863000
Powder; Molecular Formula: C12H16O7; MW: 272.26
Optical Rotation: [α]/D -64.0±2.0°, c = 3 in water
Hygroscopic
- RTECS: CE8863000
- Optical rotation: [α]/D -64.0±2.0°, c = 3 in water
- Hygroscopic
- Powder
Formula:
C₁₂H₁₆O₇ MW: 272.25 g/mol |
MDL Number:
MFCD00016915 CAS Number: 497-76-7 EINECS: 207-850-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...