Manganese ≥99.98%, granules 0.8-10 mm
Supplier: BeanTown Chemical
140055-100G
140055-25G
BT140055-100GEA
232.3
USD
BT140055-100G
BT140055-25G
Manganese ≥99.98%, granules 0.8-10 mm
Manganese
CAS: 7439-96-5; EC No: 231-105-1; MDL No: MFCD00011111; RTECS: OO9625000
Solid; Molecular Formula: Mn; MW: 54.94
Melting Point: 1244°; Boiling Point: 1962°
Density (g/mL): 7.3
- RTECS: OO9625000
- Solid
Formula:
Mn Boiling Pt: 1962 °C Melting Pt: 1244 °C Density: 7.3 g/mL |
MDL Number:
MFCD00011111 CAS Number: 7439-96-5 EINECS: 231-105-1 |
Specification Test Results
Assay | 99.98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...