Diethyl (hydroxymethyl)phosphonate ≥85%, Technical Grade
Supplier: BeanTown Chemical
139420-25G
139420-5G
BT139420-25GEA
108.57
USD
BT139420-25G
BT139420-5G
Diethyl (hydroxymethyl)phosphonate ≥85%, Technical Grade
Diethyl (hydroxymethyl)phosphonate
CAS: 3084-40-0; EC No: 221-391-6; MDL No: MFCD00014418
Liquid; Molecular Formula: C5H13O4P; MW: 168.13
Boiling Point: 124-126°/3 mmHg; Flash point:
- Refractive index: 1.444
- Liquid
Formula:
C₅H₁₃O₄P Boiling Pt: 124…126 °C/3 mmHg Density: 1.14 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00014418 CAS Number: 3084-40-0 EINECS: 221-391-6 |
Specification Test Results
Assay | 85% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...