Methyl cellulose, 4000 mPa.S (2% H₂O)

Supplier: BeanTown Chemical

Synonyms: Cellulose methyl ether

139245-100G 139245-2.5KG 139245-500G
BT139245-100GEA 212.73 USD
BT139245-100G BT139245-2.5KG BT139245-500G
Methyl cellulose, 4000 mPa.S (2% H₂O)
Methyl cellulose

CAS: 9004-67-5; MDL No: MFCD00081763 Powder


  • Powder

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR