Catechol violet

Supplier: BeanTown Chemical

138490-25G 138490-5G
BT138490-25GEA 155 USD
BT138490-25G BT138490-5G
Catechol violet
Catechol violet

CAS: 115-41-3; EC No: 204-088-3; MDL No: MFCD00005868 Powder; Molecular Formula: C19H14O7S; MW: 386.38


  • Powder

Formula: C₁₉H₁₄O₇S
MDL Number: MFCD00005868
CAS Number: 115-41-3
EINECS: 204-088-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR