(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥97%

Supplier: BeanTown Chemical

Synonyms: XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

137355-1G 137355-5G
BT137355-1GEA 66 USD
BT137355-1G BT137355-5G
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥97%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)

CAS: 161265-03-8; MDL No: MFCD00233866 Crystalline; Molecular Formula: C39H32OP2; MW: 578.62 Melting Point: 224-228°


  • Crystalline

Formula: C₃₉H₃₂OP₂
MW: 578.62 g/mol
Melting Pt: 224…228 °C
MDL Number: MFCD00233866
CAS Number: 161265-03-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR