(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: H-HoArg-OH·HCl, L(+)-Homoarginine hydrochloride, H-HomoArg-OH.HCl, H-HomoArg-OH·HCl

137160-1G 137160-25G 137160-5G
BT137160-1GEA 65 USD
BT137160-1G BT137160-25G BT137160-5G
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥98%
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride

CAS: 1483-01-8; EC No: 216-045-6; MDL No: MFCD00012622 Powder; Molecular Formula: C7H17ClN4O2; MW: 224.69 Melting Point: 213-215°


  • Powder

Formula: C₇H₁₆N₄O₂·HCl
MW: 224.7 g/mol
Melting Pt: 213…215 °C
MDL Number: MFCD00012622
CAS Number: 1483-01-8
EINECS: 216-045-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR